Learn More
Fisher Healthcare™ PROTOCOL™ Hema-G Stains for Geometric Data Hemastainer™
Quality-control tested on human blood smears to ensure proper staining and absence of precipitate
Description
Stains for manual and automated staining methods
- All PROTOCOL Hematology Stains are prepared with materials certified by the Biological Stain Commission to ensure purity
- Manufactured with strict quality control measures to ensure lot-to-lot uniformity, including extensive analytical and performance testing on human specimens
- Carefully balanced for optimum staining pH
- Remain stable for a minimum of 12 months when stored at room temperature
- Allow varied staining times to suit individual preferences
- Packaged in durable plastic bottles for safety and easy handling
- Convenient finger tabs and clear reagent identification eliminate improper line connection
Specifications
4g | |
Room Temperature | |
Hema-G Dry Buffer | |
Geometric data Hemastainer™ |
Safety and Handling
ShelfLife : 1 Year