Learn More
Fisher Healthcare™ PROTOCOL™ Stains for HEMA-TEK™ Automated Slide Stainers
Allow varied staining times to suit individual preferences
Description
Stains for manual and automated staining methods
- All PROTOCOL Hematology Stains are prepared with materials certified by the Biological Stain Commission to ensure purity
- Manufactured with strict quality control measures to ensure lot-to-lot uniformity, including extensive analytical and performance testing on human specimens
- Carefully balanced for optimum staining pH
- Remain stable for a minimum of 12 months when stored at room temperature
- Packaged in durable plastic bottles for safety and easy handling
- Convenient finger tabs and clear reagent identification eliminate improper line connection
Specifications
5 Case | |
Room Temperature | |
Liquid | |
Dark Purple |
Stain, Wright's Stain | |
37.7°C | |
Hema-Tek™ automated slide stainer |
Safety and Handling
ShelfLife : 1 Year
WARNING: Reproductive Harm - www.P65Warnings.ca.gov