Learn More
Fisher HealthCare™ Sure-Vue™ ASO Test Kit Available on GSA/VA Contract for Federal Government customers only.
Increased sensitivity ideal for testing pediatric patients
 View more versions of this product
Catalog No. 23-046-388
Due to product restrictions, please to purchase or view availability for this product.
Specifications Description & Specifications
Specifications
Non-waived
To screen both for levels of ASO in Serum greater than 200 IU/mL and levels of ASO greater than 100 IU/mL using an alternative sampling and determine titers of ASO in Serum using the semi-quantitative technique
500 Tests
Sure-Vue™ ASO Test Kit
86060, 86063
Slides (six-well), Latex reagent, positive and negative control and package insert
2 min.