Learn More
Fisher Healthcare™ Sure-Vue™ CRP Latex Test Kit
Features a suspension of latex particles coated with the IgG fraction of an anti-human, CRP-specific serum
Catalog No. 23-038001
Due to product restrictions, please to purchase or view availability for this product.This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. View substitute products »
Includes: Latex reagent, positive and negative controls, disposable slides, and package insert
This item is not returnable. View return policy
Description
Description
  • CLIA Complexity: Non-Waived
  • CPT Code: 86140/86318
  • Sensitivity: 6mg/L serum
  • Latex agglutination slide test
  • Test results in two minutes
  • Qualitative and semi-quantitative test for the determination of serum CRP, an indicator of acute infection and illness
  • For in vitro diagnostic use
Specifications
Specifications
86140, 86318
CRP-specific Antibodies in Serum
Latex reagent, positive and negative control, disposable slides and package insert
2 min.
Non-waived
Sure-Vue™ CRP Test Kit
50 Tests