Fisher HealthCare™ Sure-Vue™ CRP Latex Test Kit Available on GSA/VA Contract for Federal Government customers only.
Features a suspension of latex particles coated with the IgG fraction of an anti-human, CRP-specific serum
 View more versions of this product
Catalog No. 23-046389
    N/A
Due to product restrictions, we cannot sell this product online. Please call Customer Service at 1-800-766-7000 or send an email to CS.WebEmail@thermofisher.com for assistance. 
    Specifications
    Product Type Sure-Vue™ CRP Latex Test Kit
    CPT Code 86140, 86318
    Detectable Analytes CRP-specific Antibodies in Serum
    Includes Latex reagent, positive and negative control, disposable slides and package insert
    Quantity 500 Tests
    Test Time 2 min.
    Clia Complexity Non-waived
  • Description & Specifications
Related Products
Product Content Feedback