Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Fisher Healthcare™ Sure-Vue™ Mono Latex Test Kit
Qualitative and semi-quantitative results
Includes: Latex reagent, disposable slides (six-well), and prediluted positive and negative controls
Description
- CLIA Complexity: Non-Waived
- CPT Code: 86308
- Latex agglutination slide test designed for the qualitative and semi-quantitative detection of infectious mononucleosis heterophile antibodies in serum and plasma
- Test results in three minutes
- Sensitivity of 99% and specificity of 99.4%
- For in vitro diagnostic use
Specifications
Non-waived | |
Mononucleosis Latex Test Kit | |
1000 Tests |
Infectious Mononucleosis Heterophil Antibodies | |
Latex reagent, disposable slides (6-well) and prediluted positive and negative control |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction