Learn More
Fisher Healthcare™ Sure-Vue™ RF Rapid Latex Test Kit
Suspension of latex particles is coated with human immunoglobulin
Includes: Disposable Slides only
Description
Disposable Slides for use with Sure-Vue RF Rapid Latex Test Kit.Specifications
86430, 86318 | |
RF in Serum | |
Latex reagent, disposable slides (six-well), prediluted positive and negative control |
Non-waived | |
Disposable Slide | |
25 Pack |