Fisher HealthCare™ Sure-Vue™ RPR Test Kit Available on GSA/VA Contract for Federal Government customers only.

 View more versions of this product

Catalog No. 23-038015

    N/A

Due to product restrictions, we cannot sell this product online. Please call Customer Service at 1-800-766-7000 or send an email to CS.WebEmail@thermofisher.com for assistance. 


Includes: Dispensing pipets

    Specifications

    Description Dispensing Pipet
    Product Type Dispensing Pipet
    Detectable Analytes Detects reagin present in Serum of syphilitic patients
    For Use With (Application) For in-vitro diagnostic use
    Includes Dispensing pipets
    Quantity 500 Pack

    • CLIA Complexity: Non-Waived
    • CPT Codes: 86542/86593
    • Cardiolipin charcoal microparticle test for the qualitative and semi-quantitative determination of reagin, which is associated with syphilis
    • Antigen detects “reagin” present in serum of syphilitic patients
    • Test results in eight minutes
    • Positive results yield a flocculation of the antigen; negative results yield no reaction and a gray homogeneous suspension, or a button of non-aggregating carbon particles in the center of the circle
    • For in vitro diagnostic use

    This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your 9-digit account number, or become a registered user on fishersci.com by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service at 1-800-766-7000.

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit