Learn More
Fisher HealthCare™ Sure-Vue™ Rubella Latex Test Kit Available on GSA/VA Contract for Federal Government customers only.
Requires a dilution step for 10 IU sensitivity
 View more versions of this product
Catalog No. 23-038005
Due to product restrictions, please to purchase or view availability for this product.
Specifications Description & Specifications
Specifications
Non-waived
Rubella virus Antibodies in Serum
500 Tests
Rubella Latex Test Kit
86762, 86406
Latex reagent, disposable slides (15-well), spreading pipets, diluent Buffer, high and low positive control and negative control
8 min.