Fisher Healthcare™ Tissue-Plus™ O.C.T Compound
For in vitro diagnostic use
 View more versions of this product
Catalog No. 23-730-572
  Check Availability
Add to Cart
    Specifications
    Product Type O.C.T™ Compound
    Quantity 12 Case
    Capacity (English) 4 oz.
    Capacity (Metric) 118mL
    Capacity 4 oz., 118mL
    Color Blue
    For Use With (Application) Used as a embedding medium for frozen tissue specimens to esure optimal cutting temperature, provides a convenient specimen matrix for cryostat sectioning
  • Description & Specifications
Related Products
Product Content Feedback