Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Fisher Science Education Bio-Fresh Concentrate
Keep your specimens moist and preserved.
$17.25 - $49.00
Specifications
For Use With (Application) | Use as a humectant during dissections to keep specimens moist, flexible and mold-free |
---|
Catalog Number | Capacity (English) | Capacity (Metric) | Product Type | Quantity | Price | Quantity & Availability | |||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Capacity (English) | Capacity (Metric) | Product Type | Quantity | Price | Quantity & Availability | |||
S601010
![]() |
32 oz. | 946.3mL | Biofresh Spray Bottle | 12 Case |
Pack for $17.25
|
|
S6010S
![]() |
32 oz. | 946.3mL | Bio-fresh | 12/CS |
Pack for $18.75
|
|
S6011S
![]() |
128.4 oz. | 3797.2mL | Bio-fresh | Each |
Pack for $49.00
|
|
Description
- Use as a humectant during your dissections to keep your specimens moist, flexible and mold-free
- Use a spray bottle or atomizer to mist your specimens during use and just before sealing them at the end of class
- Dilute per directions on the bottle
- Contains no formaldehyde and is lightly scented
Specifications
Use as a humectant during dissections to keep specimens moist, flexible and mold-free |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title