Learn More
DeltaBio Fetal Pigs
SCI_ED
Sus scrofa.
Supplier:  DeltaBio 140.1189.10
 View more versions of this product
Catalog No. S1190S10
Add to Cart
Description
Description
  • Ideally suited for high school and freshman college courses where the emphasis is on basic internal anatomy
  • Perfect for the beginning biology class because of its cost, availability, size and easy dissection
  • Artiodactyla order is characterized by having an even number of toes
  • Bio-Fresh™ specimens are fixed in formaldehyde, rinsed and packaged in Bio-Fresh solution
Note: Because it is a fetal specimen, musculature is not fully toned for an in-depth dissection.
Order Info
INJECTION KEY
  • Plain: Circulatory system not injected with colors
  • Single: Arteries injected with red latex
  • Double: Arteries red, veins blue
Specifications
Specifications
Fetal Pig
Plain
35.56+ cm
Pail
Bio-Fresh™
13+ in.
10/Pk.
Product Suggestions
Product Suggestions