Learn More
Fisher Science Education™ Microscope Replacement Bulbs
SCI_ED
$14.50 - $16.25
Specifications
Type Replacement Bulb
View More Specs
Products 4
Catalog Number Mfr. No. For Use With (Equipment) Voltage Wattage Price Quantity & Availability  
Catalog Number Mfr. No. For Use With (Equipment) Voltage Wattage Price Quantity & Availability  
S74021
SCI_ED
View Documents Promotion Details
N/A Non LED model microscopes 12 V 10 w
Each for $14.50
Add to cart
S74023
SCI_ED
View Documents Promotion Details
N/A 450 LED series stereoscopic microscopes 12 V 10 w
Each for $16.25
Add to cart
S74019
SCI_ED
View Documents Promotion Details
N/A 420 Series stereoscopic microscopes 115 V 5 w This item has been discontinued and is no longer available. Please call customer service for assistance: 1-800-766-7000. N/A
S74024
SCI_ED
View Documents Promotion Details
Fisher Science Education™
800411
410 Series stereoscopic microscopes 12 V 10 w This item has been discontinued and is no longer available. Please call customer service for assistance: 1-800-766-7000. N/A
Description
Description
Specifications
Specifications
Replacement Bulb