Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Fisher Science Education™ Primary Stereomicroscopes Products for Science Education

Popular stereomicroscopes for education and industry.

$237.00

Specifications

Objective Types 2X
Eyepiece Paired widefield 10x
Lighting Type Incidental/Transmitted
View More Specs

Includes: Vinyl dustcover

Products 1
Catalog Number Magnification Power Head Style Price Quantity & Availability  
Catalog Number Magnification Power Head Style Price Quantity & Availability  
S71004A Products for Science Education
Accessories
20X Inclined head Each for $237.00
Add to cart
 
Description

Description

Rugged and dependable, delivers a 3-dimensional, unreversed image with great resolution,
brightness and clarity.

Optical System

  • Paired widefield 10x eyepieces are supplied standard, with diopter adjustment on left side
  • Interpupillary adjustment from 50mm to 75mm
  • Objectives are parfocalled, parcentered, achromatic and fixed permanently in place
  • Standard magnification of 20x is considered ideal
  • Optional WF15x eyepieces increase magnification by 50%
Illumination
  • High intensity, 12V 10W built-in incidental (top) and transmitted (bottom) illumination
  • Focusing
    • Rack and pinion focusing with slip clutch and tension adjustment
    • Mounted on metal post, adjusts up and down
    Stage
    • 80mm reversible black/white plastic contrast plate and 80mm frosted glass stage plate
    • Locked-on stage clips
    Frame
    • Cast frame, gray enamel finish

    Warranty and Services

    Five-year limited warranty, excluding bulbs and fuses

    Specifications

    Specifications

    2X
    Paired widefield 10x
    Incidental/Transmitted
    Provide Content Correction

    We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

    Product Title

    By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

    Cancel Submit