Fisher Scientific™ Accessories for PowerGen™ High Throughput Homogenizers

For use with PowerGen™ High Throughput Homogenizer

 View all options for this product

Catalog No. 02-215-518

  Check Availability
Add to Cart


    Contains 24 pins
    Type Magnet extractor (24 pin)
    For Use With (Equipment) PowerGen™ high throughput homogenizer
    Material Magnet

    • Vials are supplied with caps and are available with or without stainless-steel mixing beads
    • Magnetic pipet tips are used to remove beads from microplates
    • Bead dispenser places beads into plates easily and efficiently

  • Description & Specifications

Related Products

Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit