Fisher Scientific™ accuFlow™ Flushable Junction pH Combination Electrodes
Flushable double junction eliminates clogging
 View more versions of this product
Catalog No. 13-620-110
 N/A
N/A
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. 
    Specifications
    Connector Types BNC / Mini-Phone ATC
    pH Range 0 to 14pH
    Material Plastic
    Filling Solution Type SP138-500
    Temperature (Metric) Range 0 to 100°C
  • Description & Specifications
Related Products
Product Content Feedback