Fisher Scientific™ accumet™ AB Benchtop Meters - PROMO
Intuitive, simple operation and high accuracy in a compact, affordable meter.
 View more versions of this product
Catalog No. 13-636-AB150S
 N/A
N/A
This item has been discontinued and is no longer available. Please call customer service for assistance: 1-800-766-7000. 
    Specifications
    Model AB150 pH BioBasic Kit
    Accuracy (mV) ±0.2mV
    Voltage 100/240V
    Range (pH) -2 to 20
    Resolution (pH) 0.1/0.01/0.001
    Accuracy (pH) ±0.002
    Accuracy (Temperature) ±0.3°C
    Range (mV) ±2000mV
    Warranty Meters: Three years parts and labor; Electrodes (included with meter kits): One year parts and labor
    Includes Meter, TRIS compatible accuTupH pH electrode (13-620-183A), ATC probe (13-620-19), electrode arm, 110/220V power supply, and manual
    Display Type LCD
  • Description & Specifications
Related Products
Product Content Feedback