Fisher Scientific™ accumet™ AB15+ Basic and BioBasic™ pH/mV/°C Meters

Compact, affordable pH/mV meters with intuitive operation

 View more versions of this product

Catalog No. 13-636-AB15PC

 N/A
N/A

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.  View substitute products »


    Specifications

    Display Type LCD
    Includes accumet liquid-filled, glass body single-junction combination pH Ag/AgCl Electrode (13-620-285), separate ATC probe (13-620-19)
    Accuracy ±0.002pH ±1 least significant digit
    Detection Range -2 to +20pH
    Electrical Requirements 115V 60Hz
    For Use With (Application) pH, mV, Temperature
    Hertz 60Hz
    Resolution 0.1/0.01/0.001pH
    Voltage 115V

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit