Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Fisherbrand accumet AB15+ Basic and BioBasic pH/mV/°C Meters
Compact, affordable pH/mV meters with intuitive operation
Specifications
Electrical Requirements | 120V, 50/60Hz |
---|---|
For Use With (Application) | pH, mV, Temperature |
Frequency | 60 Hz |
Voltage | 120 V |
Display Type | LCD |
Includes: Meter Only package includes meter, power supply, electrode support (13-637-671) and operation manual.
Meter Kit package includes items above plus an accumet liquid-filled, glass body single-junction combination pH Ag/AgCl Electrode (13-620-285) and separate ATC probe (13-620-19).
BioBasic Meter Kit package for the life science lab includes meter, power supply, manual, electrode support and accuTupH rugged bulb, liquid-filled, glass body double-junction combination pH Combination Electrode (13-620-183) and separate ATC probe (13-620-19).
Note: Each package is also available with a Certificate of Traceability to NIST Standards.
Catalog Number | Includes | Price | Quantity & Availability | ||||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Includes | Price | Quantity & Availability | ||||||
13-636-AB15PB
|
Power supply, electrode support and accuTupH rugged bulb, double-junction combination pH electrode (13-620-183), separate ATC probe |
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. |
View substitute products » |
Description
- Intuitive operation and improved user interface Large LCD guides user with plain-language prompts and error messages
- Display indicators keep you apprised of menu choice, measuring units, buffers standardized (in pH mode) and temperature
- No cryptic icons to learn: prompts and status/error messages are displayed in plain language
- Store up to 100 datapoints in nonvolatile memory
- Selectable resolution to 0.1, 0.01 or 0.001pH
- Five-button, soft-key input simplifies data entry
- Internal diagnostics
- Automatic recognition of 15 buffers and five-point calibration
- BNC and reference pin inputs, input for accuFet Electrode, ATC probe input, plus bidirectional RS-232 port
Order Info
Dimensions: 7.5L x 5.5W x 3.25 in.H (19 x 14 x 8cm). Net weight: 1.875 lb. (0.8kg).
Warranty and Services
accumet Model AB15+ is warranted to the end user to be free from defects in materials and workmanship under normal use for a period of three years.
Specifications
120V, 50/60Hz | |
60 Hz | |
LCD |
pH, mV, Temperature | |
120 V |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title