Learn More
Fisher Scientific™ accumet™ AB15 Basic and BioBasic™ pH/mV/°C Meters
Simple, menu-guided operation in an affordable pH/mV meter
 View more versions of this product
Catalog No. 13-636-AB15B
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
View substitute products »
Specifications Description & Specifications
Specifications
Double-junction combination pH electrode 13-620-183, ATC Probe 13-620-19, electrode support arm and bracket, manual, power supply
±0.002pH ±1 least significant digit
115V 60Hz
60Hz
115V
LCD
-2 to +20pH
pH, mV, Temperature
0.1/0.01/0.001pH