Learn More
Fisher Scientific™ accumet™ AB15+ Basic and BioBasic™ pH/mV/°C Meters
Compact, affordable pH/mV meters with intuitive operation
 View more versions of this product
Catalog No. 13-636-AB15P
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
View substitute products »
Specifications Description & Specifications
Specifications
Liquid-filled, glass body single-junction combination pH Ag/AgCl electrode (13-620-285), separate ATC probe (13-620-19)
Multiple
115V 60Hz
60Hz
115V
LCD
Multiple
pH, mV, Temperature
Multiple
Accessories