Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Fisherbrand accumet Excel XL15 pH/mV/Temperature Meter
Accurate measurements, easy-to-use touchscreen design
Specifications
Electrical Requirements | 120/240V, 50/60Hz |
---|---|
Frequency | 50/60 Hz |
Voltage | 120/240 V |
Display Type | LCD |
Includes: Meter Only package includes meter, electrode support arm and bracket, 110/220V power supply and operation manual. Also available with Certificate of NIST Traceability
Meter Kit/pH package includes meter, accuCap combination pH electrode (13-620-130), ATC probe (13-620-19), electrode support arm and bracket, 110/220V power supply and operation manual. Also available with Certificate of NIST Traceability.
Each package aboveis also available with Certificate of Traceability to NIST Standards.
Meter/Software Kit package includes meter, accuCap combination pH electrode (13-620-130), ATC probe (13-620-19), electrode support arm and bracket, 110/220V power supplies and operation manual, plus powerful, comprehensive data management software.
Catalog Number | Description | Price | Qty | Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Description | Price | Qty | Availability | |||||
13-636-XL15
|
N/A | Excel XL15 pH Meter Kit |
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. |
View substitute products » |
Description
Simple but powerful; large, full color, touchscreen display
- Auto-calibrates with up to six pH buffers from three standard sets and 15 different buffers; custom buffers and manual calibration pH mode accepts standard or accuFet nonglass electrodes
- Windows CE-driven, full-color touchscreen provides unmatched ease of use in operation and setup
- Windows CE operating system makes operation intuitive from the first time you turn it on Easily recognizable icons help you get to setup or measurement modes quickly, for both everyday measurements and more complicated routines
- Extended pH range: -2 to +20pH; reads to 0.1mV for accurate titrations and ORP readings, too
- Adjustable resolution to 0.001pH Calibration at up to six points for exceptional accuracy of ±0.002pH
- Customizable printer/computer output
- 1000-reading datalog capability
- Print Setup lets you print exactly what you want, when you want
- Displays time in 12- or 24-hour format, date in U.S. or international format
- USB port lets you connect keyboard, mouse, or trackball for meter control, or connect printer or PC for data output
- RS-232 serial port lets you connect to PCs or printers without USB, or frees computer USB port while using the meter is in use
- Ethernet allows unmatched LAN connectivity and even allows software upgrades
- Designed in compliance with FDA's 21 CFR Part 11 regulations
- This software lets you expand the memory space of your meter to your personal computer's hard disk, and manage multiple projects under separate ID logins
- Password security protects each ID login's data from tampering and accidental deletion.
Order Info
- Durable ABS plastic housing resists dust and chemicals
- Dimensions: 9.25L x 6.5W x 3.5 in.H (23.5 x 16.5 x 9cm)
- Net weight 2.75 lb. (1.2kg)
Warranty and Services
accumet Three-Year Warranty. accumet Model XL15 is warranted to the end user to be free from defects in materials and workmanship under normal use for a period of three years. Call your Fisher Scientific Customer Service Representative for full details.
Specifications
120/240V, 50/60Hz | |
50/60 Hz | |
120/240 V | |
LCD |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title