Fisher Scientific™ accumet™ Excel XL15 pH/mV/Temperature Meter

Accurate measurements, easy-to-use touchscreen design

 View more versions of this product

Catalog No. 13-636-XL15

 N/A
N/A

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.  View substitute products »


    Specifications

    Display Type LCD
    Includes Meter, accuCap combination pH electrode, ATC probe, electrode support, manual
    Accuracy Multiple
    Detection Range Multiple
    Electrical Requirements 110 to 220V 50/60Hz
    For Use With (Application) pH, mV, Temperature
    Resolution Multiple
    Voltage 110 to 220V
    Hertz 50/60Hz

  • Description & Specifications
  • Accessories

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit