Learn More
accumet™ Gas-Sensing Combination ISEs Available on GSA/VA Contract for Federal Government customers only.
For ammonia and carbon dioxide analyses; tough, chemical-resistant body; permanently bonded membrane/cap assembly—no difficult user assembly required
 View more versions of this product
Catalog No. 13-620-508
  Check Availability
Add to cart
Specifications Description & Specifications
Specifications
Determines Ammonia and Ammonium ion concentration in sewage effluent, boiler water, industrial wastes, stack gases, food, fertilizers
0.5 in.
Noncrystallizing - NH4Cl
102mm
0.009 to 1700ppm
0° to 50°C
13
<300megohm
0 to 50°C
U.S. Standard/Pin
13mm
4.01 in.
Epoxy Body
32° to 122°F
ASTM, EPA
Volatile Amines, Metal cations that complex Ammonia
<30 sec.
Accessories