Learn More
Fisher Scientific™ accumet™ Glass Body Ag/AgCl Reference Electrodes - Mercury-Free Available on GSA/VA Contract for Federal Government customers only.
Recommended for stability at higher temperatures and for use with strongly basic samples
 View more versions of this product
Catalog No. 13-620-45
  Check Availability
Add to cart
Specifications Description & Specifications
Specifications
Recommended for stability at higher temperatures and for use with strongly basic samples
Pin
Ag/AgCl
Glass
32° to 212°F
SP138-500
Single-Junction High Temperature Ag/AgCl Reference with Side Arm
0° to +100°C
4mol/L KCl with AgCl
Single
0 to 14
0° to +100°C
Suggested Indicating Electrodes: 13-620-1, 13-620-295, 13-620-284, 13-620-3, 13-620-186, 13-620-187, 13-620-115