Fisher Scientific™ accumet™ Glass Body Calomel Reference Electrodes Available on GSA/VA Contract for Federal Government customers only.

 View more versions of this product

Catalog No. 13-620-57

  Check Availability
Add to Cart

Alert

The products indicated contain mercury. Don't put products containing mercury in the trash. Products containing mercury are to be recycled or disposed of as hazardous waste. Products containing mercury may be regulated by Local, State, or Federal Laws. Some products containing mercury are not available for sale in some states.


    Specifications

    Length (Metric) Cable 76.2cm
    Material Glass
    pH Range 0 to 14
    Temperature (Metric) Operating -5° to +80°C
    Connector Types Pin
    Diameter (English) Body 0.37 in.
    Internal Reference Hg/Hg2Cl2 (Calomel)
    Junction Cracked Bead
    Temperature (Metric) Range -5 to +80°C
    Applications Biological, Acid, Nonaqueous Solutions
    Length (English) 5.75 in.
    Length (English) Cable 30 in.
    Length (Metric) 14.6cm
    Temperature (English) Operating 23° to 176°F
    Reference Electrolyte Calomel
    Diameter (Metric) Body 1cm
    Diameter (Metric) Cap 1.6cm
    Filling Solution Type Mercury

  • Description & Specifications
  • Accessories

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit