Learn More
Fisher Scientific™ accumet™ Glass Body Calomel Reference Electrodes
 View more versions of this product
Catalog No. 13-620-61
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
View substitute products »
Alert:
The products indicated contain mercury. Don't put products containing mercury in the trash. Products containing mercury are to be recycled or disposed of as hazardous waste. Products containing mercury may be regulated by Local, State, or Federal Laws. Some products containing mercury are not available for sale in some states.
Specifications Description & Specifications
Specifications
Emulsions, Low Ionic Strength Solutions, Slurries, Suspensions
Pin
1cm
Mercury
Reverse sleeve
30 in.
76.2cm
0 to 14
-5° to +80°C
-5 to +80°C
0.37 in.
1.6cm
Hg/Hg2Cl2 (Calomel)
5.75 in.
14.6cm
Glass
23° to 176°F
Calomel
Accessories