Learn More
Fisher Scientific™ accumet™ Glass Body Mercury-Free Reference Electrodes
Mercury-free, refillable, TRIS-compatible reference electrodes
 View more versions of this product
Catalog No. 13-620-857
  Check Availability
Add to cart
Specifications Description & Specifications
Specifications
Biomedical, TRIS
0.37 in.
1.4cm
30 in.
76.2cm
0 to 14
-5° to +80°C
Pin
1cm
2.5 in.
6.35cm
Glass
23° to 176°F