Learn More
Fisher Scientific™ accumet™ Liquid-Filled Polymer Body Combination Electrodes
Ideal for measurements in small vessels or with small samples
 View more versions of this product
Catalog No. 13-620-300
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
View substitute products »
Specifications Description & Specifications
Specifications
-5 to +80°C
Polypropylene
SP138-500
10mm
Calomel or Ag/AgCl
Ideal for handheld use with aqueous samples
pH combination electrode
0 to 14
Single Ceramic
106mm
BNC
4 in.
0.4 in.