Learn More
Fisher Scientific™ accumet™ Micro and Semimicro Size Glass Combination Electrodes - Mercury - Free Available on GSA/VA Contract for Federal Government customers only.
Ideal for microwell plates, serum cups, cuvets, micro test tubes, NMR tubes and capillary tubes
 View more versions of this product
Catalog No. 13-620-94
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Specifications
Description
Ideal for microwell plates, serum cups, cuvets, micro test tubes, NMR tubes and capillary tubes
Description & Specifications
Specifications
-5° to +110°C
0 to 14
Glass
Saturated KCl with AgCl. KCl
U.S. Standard
23° to 230°F
Micro
1 Year
-5° to 110°C
Single
60mm
Ag/AgCl
Small samples
pH/ORP combination titration electrode, Miniature Ag/AgCl
Accessories