Fisher Scientific™ accumet™ pH Indicating Glass Body Universal Glass Membrane Models- Mercury-Free
 View more versions of this product
Catalog No. 13-620-853
  Check Availability
Add to Cart
    Specifications
    Material Glass
    pH Range 0 to 14
    Connector Types BNC
    Length (English) 30 in.
    Electrode Best for use with AgAgCl (non-calomel) reference half cells
  • Description & Specifications
Related Products
Product Content Feedback