Learn More
Fisher Scientific™ accumet™ Solid State Combination ISEs, Mercury-Free Available on GSA/VA Contract for Federal Government customers only.
For bromide, chloride, copper, cyanide, fluoride, lead and silver/sulfide determinations
 View more versions of this product
Catalog No. 13-620-528
  Check Availability
Add to cart
Specifications Description & Specifications
Specifications
Used in analyses of water, wastewater, stack gases, explosives, etching baths, research with biological fluids and tissues, beverages, detergents
0.5 in.
SP138-500
108mm
0.02 to saturationppm
0° to 80°C
5 to 8
Fluoride Combination
U.S. Standard/Pin
13mm
4.25 in.
Glass Body
32° to 176°F
ASTM, EPA
OH-
0 to 80°C
Accessories