Fisher Scientific™ accumet™ Solid-State Half-Cell ISEs - Mercury - Free Available on GSA/VA Contract for Federal Government customers only.
For bromide, chloride, fluoride, lead and silver/sulfide determinations
 View more versions of this product
Catalog No. 13-620-521
  Check Availability
Add to Cart
    Specifications
    Applications Determination of Bromide concentration in aqueous solutions, certain nonaqueous solutions, biological fluids, soil, plants, food, water, effluents
    Material Epoxy
    pH Range 0 to 14
    Connector Types BNC
  • Description & Specifications
Related Products
Product Content Feedback