Fisher Scientific™ accumet™ Solid-State Half-Cell ISEs - Mercury - Free Available on GSA/VA Contract for Federal Government customers only.
For bromide, chloride, fluoride, lead and silver/sulfide determinations
 View more versions of this product
Catalog No. 13-620-522
  Check Availability
Add to Cart
    Specifications
    Applications Determination of Fluoride concentration in water/wastewater, stack gases, explosives, etching baths, bioresearch, beverages, detergents
    Material Epoxy
    pH Range 5 to 9
    Connector Types U.S. Standard
  • Description & Specifications
Related Products
Product Content Feedback