Learn More
Fisher Scientific™ accumet™ Solid-State Half-Cell ISEs - Mercury - Free Available on GSA/VA Contract for Federal Government customers only.
For bromide, chloride, fluoride, lead and silver/sulfide determinations
 View more versions of this product
Catalog No. 13-620-545
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Specifications Description & Specifications
Specifications
Determination of Silver/Sulfide concentration in aqueous solutions, certain nonaqueous solutions, water, pulping liquors
BNC
Epoxy
2 to 12
Accessories