Learn More
Fisherbrand accumet Solid-State Half-Cell ISEs - Mercury - Free
For bromide, chloride, fluoride, lead and silver/sulfide determinations
$731.00
Catalog Number | For Use With (Application) | Connector Types | pH Range | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | For Use With (Application) | Connector Types | pH Range | Price | Quantity & Availability | ||||
13-620-523
|
Determination of Fluoride concentration in water/wastewater, stack gases, explosives, etching baths, bioresearch, beverages, detergents | BNC | 5 to 9 |
Each for $731.00
|
|
13-620-519
|
Determination of Chloride concentration in water/wastewater, soil, dairy and tomato based food products, meats, vegetables and in research with biomedical fluids | BNC | 0 to 14 |
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. |
View substitute products » |
13-620-522
|
Determination of Fluoride concentration in water/wastewater, stack gases, explosives, etching baths, bioresearch, beverages, detergents | U.S. Standard | 5 to 9 |
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. |
View substitute products » |
Description
For bromide, chloride, fluoride, lead and silver/sulfide determinations
- pH range: 5-9
- Material: Epoxy
- Connector type: BNC
Specifications
Epoxy |