Fisher Scientific™ accumet™ Solid-State Half-Cell ISEs - Mercury - Free
For bromide, chloride, fluoride, lead and silver/sulfide determinations
 View more versions of this product
Catalog No. 13-620-519
  Check Availability
Add to Cart
    Specifications
    Applications Determination of Chloride concentration in water/wastewater, soil, dairy and tomato based food products, meats, vegetables and in research with biomedical fluids
    Material Epoxy
    pH Range 0 to 14
    Connector Types BNC
  • Description & Specifications
Related Products
Product Content Feedback