Learn More
Fisher Scientific™ accumet™ Universal pH/ATC Electrodes
Combination pH mercury-free electrodes with built-in temperature compensation
 View more versions of this product
Catalog No. 13-621-703
  Check Availability
Add to cart
Specifications Description & Specifications
Specifications
0° to 80°C
Epoxy
Gel
WTW, Pinnacle (part numbers ending in P) meter
Cable with BNC and ATC connectors, electrode storage bottle
0° to 80°C
Double
Banana
32° to 176°F
pH/ATC combination electrode