Fisher Scientific™ accumet™ Waterproof AP84 Portable pH/Dissolved Oxygen Meter Available on GSA/VA Contract for Federal Government customers only.

Wide-range pH and rapid DO readings

 View more versions of this product

Catalog No. 13-636-AP84

 N/A
N/A

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.  View substitute products »


    Specifications

    Product Type AP84 Portable Meter Kit
    Warranty 3 Years
    Resolution Multiple
    Resolution (Temperature) 0.1°C
    Resolution (pH) 0.01
    Accuracy Multiple
    Accuracy (DO) ±1.5% FS
    Calibration Points (pH) 5 Points (pH 1.68, 4.01, 7, 10, 12.45)
    Length (Metric) 19cm
    Range Multiple
    Temperature (English) Range 32° to 122°F
    Temperature (Metric) Range 0° to 50°C
    Width (English) 3.75 in.
    Width (Metric) 9.5cm
    Includes Meter, pH electrode, DO/temperature probe, pH buffer solutions, electrolyte solution, two replacement membrane caps, 50mL each pH 4 and pH 7 standard solutions, rinse bottle, instructions, four AAA batteries and carrying case
    Range (pH) -2 to 16
    Calibration Points 5
    Accuracy (Temperature) ±0.3°C
    Display Type LCD
    Accuracy (pH) ±0.01
    Battery Type AAA
    For Use With (Application) pH, DO, Temperature
    Height (English) 2.25 in.

  • Description & Specifications
  • Accessories

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit