Fisher Scientific™ accumet™ XL500 Dual pH/ISE and Conductivity Benchtop Meters Available on GSA/VA Contract for Federal Government customers only.

Provide advanced measurements with simple operation and offer a large, vivid color touch-screen display that allows for dynamic functionality. Fisher Scientific accumet XL500 dual pH/ISE/conductivity benchtop meters offer advanced features with simple operation to help ensure your measurements are reliable and your laboratory processes are efficient.

 View more versions of this product

Catalog No. 13-636-XL500

  Check Availability
Add to Cart

Includes: accumet XL500 dual pH/ISE & conductivity benchtop meter, TRIS-compatible double junction refillable glass-body pH electrode (13-620-183A), ATC probe (13-620-19), 2-cell conductivity/temperature probe (13-620-100), electrode stand, RS232 and USB cables, 100/240V universal power supply

    Specifications

    Hertz 50/60Hz
    Accuracy (ISE) 0.5 % full scale (monovalent ion), 1 % full scale (divalent ion)
    Accuracy (TDS) ±1 % full scale
    Accuracy (Salinity) ±1 % full scale
    Accuracy (Resistivity) ±1 % full scale
    Dimensions (L x W x H) 24.6 x 17.3 x 9.9cm (9.7 x 6.8 x 3.9 in.)
    Warranty 3 Years
    Height (English) 3.89 in.
    Length (Metric) 24.6cm
    Resolution Multiple
    Range (ISE) ±2000mV
    Width (Metric) 17.3cm
    Range (TDS) 0.050ppm to 500ppt (at TDS factor 1.00)
    Resolution (Salinity) 0.01/0.1ppm, 0.001/0.01/0.1ppt
    Range (Resistivity) 3W to 100MW over 4 ranges: 0 to 20kW, 20 to 200kW, 200kW to 2MW, 2 to 100MW
    Resolution (Resistivity) 0.01/0.1W, 0.001/0.1kW, 0.01MW
    Range (pH) -2 to 20
    Resolution (pH) 0.1/0.01/0.001
    Resolution (Temperature) 0.1°C
    Data Points 2000
    Inputs 2 BNC, 2 ATC
    Weight (Metric) 1.13kg
    Weight (English) 2.5 lb.
    Detection Range Multiple
    Height (Metric) 9.9cm
    Length (English) 9.68 in.
    Model XL500 Dual pH/ISE/Conductivity Kit
    Accuracy (mV) ±0.2mV
    Voltage 100/240V
    Resolution (ISE) 2/3/4 Digits
    Range (Conductivity) 0 to 500mS
    Resolution (Conductivity) 0.001/0.01/0.1mS
    Accuracy (Conductivity) ±1% FS
    Resolution (TDS) 0.01/0.1ppm, 0.001/0.01/0.1ppt
    Range (Salinity) 0 to 80ppt
    Includes Meter, TRIS compatible accuTupH pH electrode (13-620-183A), ATC probe (13-620-19), Conductivity/Temp. probe (13-620-100), electrode arm, RS-232 and USB cables, and 110/220V power supply
    Cell Constant 0.01 to 10
    Calibration Points (pH) Up to 6 pre-set or 5 custom
    Calibration Points (ISE) 2 to 6
    Calibration Points (Conductivity) Automatic (4 points), maximum 1/range, manual (5 points), maximum 1/range
    Accuracy (pH) ±0.002
    Accuracy (Temperature) ±0.3°C
    Display Type LCD
    Range (mV) ±2000mV
    Data Management Details 3 to 86400 sec.
    Outputs RS232, USB-A, Mini-B USB, RJ45, stirrer port (2)
    Certifications/Compliance NIST, ISO, FDA, USDA, EPA
    Accuracy Multiple
    Resolution (mV) 0.1mV

    • High resolution color display shows data clearly from wide viewing angles and distances
    • Optimized touch-screen with glass/glass sensitivity provides great ease of use
    • Onscreen info includes date and time, stability indicator, alarms and context-specific “help” screens
    • One to six point pH calibration with 0.001 pH resolution
    • Automatic buffer recognition with six pH buffer sets or custom buffer option
    • Two to six point ISE calibration with 0.001 ppm, mg/L or molar resolution
    • ISE methods for known addition, known subtraction, analyte addition and analyte subtraction
    • Conductivity, TDS (total dissolved solids), salinity and resistivity modes
    • Automatic or manual temperature compensation, displayed in °C or °F
    • Display up to three measurements simultaneously (two pH or ISE, one conductivity)
    • Up to 10 password protected user ID accounts, each retains unique calibration and instrument settings with memory to store 2000 data points
    • RS232 and USB interfaces for printer or computer interfacing and real-time onscreen graphing of measurements
    • RJ45 Ethernet/Internet port to connect Local Area Network (LAN) or Internet
    • Bidirectional communication for LIMS connectivity and 3rd party software
    • Connectivity for peripherals such as keyboard, mouse or other USB device
    • Multilanguage interface with English, German, Chinese, French, Italian and Spanish
    • Two ports for optional stirring probe (Cat. No. 13-620-BSP) with five speed settings
    • Option to wall-mount meter to create additional bench space
    NIST-Traceable Calibration Models (-CERT)
    • Help meet ISO, FDA, USDA and EPA quality assurance guidelines
    • Includes meter accuracy at five test points across pH range
    • Includes meter accuracy at five test points across corresponding mV range
    • Testing performed by accredited laboratory, includes NIST-Traceable certificate
    Specifications
    • pH Range: -2.000 to 20.000 pH
    • pH Resolution: 0.1, 0.01, 0.001 pH
    • pH Accuracy: ±0.002 pH
    • ISE Range: 0.001 to 19999 ppm, mg/L or molar
    • ISE Resolution: 0.001, 0.01, 0.1, 1 (automatic)
    • ISE Accuracy: 0.5 % full scale (monovalent ion); 1 % full scale (divalent ion)
    • mV Range: ±2000.0mV
    • mV Resolution: 0.1mV
    • mV Accuracy: ±0.2mV or ±0.05 %, whichever is greater
    • mV Offset Adjustment: Up to ±150mV
    • Conductivity Range: 0.00µS to 500.0mS
    • Conductivity Resolution: 0.01, 0.1µS; 0.001, 0.01, 0.1mS
    • Conductivity Accuracy: ±1 % full scale
    • TDS Range: 0.00ppm to 500 ppt (TDS factor 1.00)
    • TDS Resolution: 0.01, 0.1 ppm; 0.001, 0.01, 0.1 ppt
    • TDS Accuracy: ±1 % full scale
    • Salinity Range: 0 to 80.0 ppt
    • Salinity Resolution: 0.01, 0.1 ppm; 0.001, 0.01, 0.1 ppt
    • Salinity Accuracy: ±1 % full scale
    • Resistivity Range: 2.000 Ohm to 20.0 MOhm
    • Resistivity Resolution: 0.01, 0.1 Ohm; 0.001, 0.1 kOhm; 0.01 MOhm
    • Resistivity Accuracy: ±1 % full scale
    • Temperature Range: -5° to +105°C (23° to 221.0°F)
    • Temperature Resolution: 0.1°C (0.1°F)
    • Temperature Accuracy: ±0.2°C (±0.3°F)
    • Calibration Points: 1 to 6 points with automatic buffer recognition (ABR)
    • ISE Calibration Points: 2 to 6 points
    • ATC Temperature Calibration: Offset in 0.1°C increments; Offset range: ±5°C (±9°F)
    • Memory: 2000 data sets per user ID with date/time stamp
    • Self-Test: Yes
    • Inputs: Two BNC, 2 mini-phone ATC, 8 Pin DIN (conductivity), ground
    • Output: RJ45, RS232, 2 USB, two stirring probe
    • Power Requirements: 100 to 240VAC, 50/60Hz
    • Dimensions (L x W x H): 24.6 x 17.3 x 9.9cm (9.7 x 6.8 x 3.9 in.)
    • Weight: 1.13kg (2.5 lb.)
    Applications
    Pharmaceutical, biotech, research, QA/QC, industrial, food, beverage

    Certifications

    CE, TUV 3-1, FCC Class A

    Warranty

    3 years on meter

  • Description & Specifications
  • Promotions

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit