Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Fisherbrand™ accumet™ XL500 Benchtop Dual pH/ISE and Conductivity Meters

Provide advanced measurements with simple operation and offer a large, vivid color touch-screen display that allows for dynamic functionality. Fisherbrand accumet XL500 dual pH/ISE/conductivity benchtop meters offer advanced features with simple operation to help ensure your measurements are reliable and your laboratory processes are efficient.

 View more versions of this product

Catalog No. 13-636-XL500A


Add to Cart

Includes: accumet XL500 dual pH/ISE & conductivity benchtop meter, electrode stand, RS232 and USB cables, 100/240V universal power supply

Description

Description

Provide advanced measurements with simple operation and offer a large, vivid color touch-screen display that allows for dynamic functionality. Fisherbrand accumet XL500 dual pH/ISE/conductivity benchtop meters offer advanced features with simple operation to help ensure your measurements are reliable and your laboratory processes are efficient.

  • High resolution color display shows data clearly from wide viewing angles and distances
  • Optimized touch-screen with glass/glass sensitivity provides great ease of use
  • Onscreen info includes date and time, stability indicator, alarms and context-specific “help” screens
  • One to six point pH calibration with 0.001 pH resolution
  • Automatic buffer recognition with six pH buffer sets or custom buffer option
  • Two to six point ISE calibration with 0.001 ppm, mg/L or molar resolution
  • ISE methods for known addition, known subtraction, analyte addition and analyte subtraction
  • Conductivity, TDS (total dissolved solids), salinity and resistivity modes
  • Automatic or manual temperature compensation, displayed in °C or °F
  • Display up to three measurements simultaneously (two pH or ISE, one conductivity)
  • Up to 10 password protected user ID accounts, each retains unique calibration and instrument settings with memory to store 2000 data points
  • RS232 and USB interfaces for printer or computer interfacing and real-time onscreen graphing of measurements
  • RJ45 Ethernet/Internet port to connect Local Area Network (LAN) or Internet
  • Bidirectional communication for LIMS connectivity and 3rd party software
  • Connectivity for peripherals such as keyboard, mouse or other USB device
  • Multilanguage interface with English, German, Chinese, French, Italian and Spanish
  • Two ports for optional stirring probe (Cat. No. 13-620-BSP) with five speed settings
  • Option to wall-mount meter to create additional bench space
NIST-Traceable Calibration Models (-CERT)
  • Help meet ISO, FDA, USDA and EPA quality assurance guidelines
  • Includes meter accuracy at five test points across pH range
  • Includes meter accuracy at five test points across corresponding mV range
  • Testing performed by accredited laboratory, includes NIST-Traceable certificate
Specifications
  • pH Range: -2.000 to 20.000 pH
  • pH Resolution: 0.1, 0.01, 0.001 pH
  • pH Accuracy: ±0.002 pH
  • ISE Range: 0.001 to 19999 ppm, mg/L or molar
  • ISE Resolution: 0.001, 0.01, 0.1, 1 (automatic)
  • ISE Accuracy: 0.5 % full scale (monovalent ion); 1 % full scale (divalent ion)
  • mV Range: ±2000.0mV
  • mV Resolution: 0.1mV
  • mV Accuracy: ±0.2mV or ±0.05 %, whichever is greater
  • mV Offset Adjustment: Up to ±150mV
  • Conductivity Range: 0.00μS to 500.0mS
  • Conductivity Resolution: 0.01, 0.1μS; 0.001, 0.01, 0.1mS
  • Conductivity Accuracy: ±1 % full scale
  • TDS Range: 0.00ppm to 500 ppt (TDS factor 1.00)
  • TDS Resolution: 0.01, 0.1 ppm; 0.001, 0.01, 0.1 ppt
  • TDS Accuracy: ±1 % full scale
  • Salinity Range: 0 to 80.0 ppt
  • Salinity Resolution: 0.01, 0.1 ppm; 0.001, 0.01, 0.1 ppt
  • Salinity Accuracy: ±1 % full scale
  • Resistivity Range: 2.000 Ohm to 20.0 MOhm
  • Resistivity Resolution: 0.01, 0.1 Ohm; 0.001, 0.1 kOhm; 0.01 MOhm
  • Resistivity Accuracy: ±1 % full scale
  • Temperature Range: -5° to +105°C (23° to 221.0°F)
  • Temperature Resolution: 0.1°C (0.1°F)
  • Temperature Accuracy: ±0.2°C (±0.3°F)
  • Calibration Points: 1 to 6 points with automatic buffer recognition (ABR)
  • ISE Calibration Points: 2 to 6 points
  • ATC Temperature Calibration: Offset in 0.1°C increments; Offset range: ±5°C (±9°F)
  • Memory: 2000 data sets per user ID with date/time stamp
  • Self-Test: Yes
  • Inputs: Two BNC, 2 mini-phone ATC, 8 Pin DIN (conductivity), ground
  • Output: RJ45, RS232, 2 USB, two stirring probe
  • Power Requirements: 100 to 240VAC, 50/60Hz
  • Dimensions (L x W x H): 24.6 x 17.3 x 9.9cm (9.7 x 6.8 x 3.9 in.)
  • Weight: 1.13kg (2.5 lb.)
Applications
Pharmaceutical, biotech, research, QA/QC, industrial, food, beverage

Certifications

CE, TUV 3-1, FCC Class A

Warranty and Services

3 years on meter

TRUSTED_SUSTAINABILITY
Specifications

Specifications

RS232, USB-A, Mini-B USB, RJ45, stirrer port (2)
±1% FS
±0.2 mV
± 1% full scale
± 1% full scale
2 to 6
0.01 to 10
3 to 86400 sec.
Multiple
LCD
0 to 500 mS
-2 to 20
0 to 80 ppt
2/3/4 Digits
-5°C to +105°C
2 BNC, 2 ATC
±2000 mV
0.001/0.01/0.1mS
0.01/0.1W, 0.001/0.1kW, 0.01MW
0.01/0.1ppm, 0.001/0.01/0.1ppt
0.1°C
24.6 x 17.3 x 9.9cm (9.7 x 6.8 x 3.9 in.)
24.6 cm
3.89 in.
2.5 lb.
100/240V
Multiple
0.5% full scale (monovalent ion), 1% full scale (divalent ion)
±0.002
± 1% full scale
Automatic (4 points), maximum 1/range, manual (5 points), maximum 1/range
Up to 6 preset or 5 custom
NIST, ISO, FDA, USDA, EPA
2000
2
pH, mV, ISE, Ion, Concentration, Conductivity, TDS, Salinity, Resistivity, Temperature
±2000 mV
3 W to 100 MW over 4 ranges: 0 to 20 kW, 20 to 200 kW, 200 kW to 2 MW, 2 to 100 MW
0.050 ppm to 500 ppt (at TDS factor 1.00)
0.1/0.01/0.001
Meter, electrode arm, RS-232 and USB cables, and 110/220V power supply
XL500 Dual pH/ISE/Conductivity Meter
Multiple
0.1mV
0.01/0.1ppm, 0.001/0.01/0.1ppt
±0.3°C
3 Years
9.68 in.
17.3 cm
9.9 cm
1.13 kg
50/60Hz
Product Suggestions

Product Suggestions

SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit