Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Fisherbrand™ accuSkan™ FC Filter-Based Microplate Photometer

For research and routine ELISA applications

 View more versions of this product

Catalog No. 14-377-576


Only null left
Add to Cart
Request a Quote

Description

Description

A proven ELISA solution—accuSkan FC and the Fisher Scientific accuWash washer. The combination of the accuSkan FC microplate photometer and the accu-Wash™ microplate washer offers a convenient modular solution for routine and research ELISA applications.

A proven ELISA solution—accuSkan FC and the Fisher Scientific accuWash washer. The combination of the accuSkan FC microplate photometer and the accu-Wash™ microplate washer offers a convenient modular solution for routine and research ELISA applications.
  • Broad wavelength range of 340 to 850nm for a wide variety of applications
  • Eight-position filter wheel with three standard pre-installed filters (405, 450 and 620nm)
  • Fast and accurate measurement of both 96- and 384-well plates for various throughput requirements
  • On-board linear shaking and optional incubation up to 50°C for temperature critical assays
  • Superior usability with both internal software and dedicated PC software
  • Data export capability via USB memory stick for easy data transfer
Applications: Immunoassays (ELISA), protein and enzyme assays,cytotoxicity and proliferation assays, growth curves and endotoxins

Warranty and Services

1 Year

TRUSTED_SUSTAINABILITY
Specifications

Specifications

Photodetector
Ambient +4° to 40°C
Quartz Halogen Lamp
96 and 384-well Plates
±1% (0 to 3 Abs) or ±0.003 Abs, which ever is greater at 405 nm
100/240 V, 50/60 Hz
Robotic automation
CV ≤ 0.2% (0.3 to 3 Abs) at 405 nm
1 Year
11.4 x 15.7 x 8.7 in. (29 x 40 x 22 cm)
PC Control, Internal Software
96 wells: 6 sec. (fast mode), 12 sec. (normal mode)
Linear shaking with 3 modes
accuSkan FC with incubator
8-position filter wheel; filters installed: 405nm, 450nm, and 620nm
96-well plate: 0 to 3 Abs (±2% at 405 nm)
384-well plate: 0 to 2.5 Abs (±2% at 405 nm)
0.001 Abs
340 to 850 nm
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

WARNING: Cancer - www.P65Warnings.ca.gov
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.