Learn More
Fisherbrand™ accuTupH Plus™ Rugged Bulb Variable Temperature pH Combination Electrode
Can be optimal for lab use and precise Q.C. measurements.
$528.50
Specifications
Type pH Combination Electrode
View More Specs
Products 1
Catalog Number Connector Types Filling Solution Type Price Quantity & Availability  
Catalog Number Connector Types Filling Solution Type Price Quantity & Availability  
13-620-185
Accessories View Documents
BNC Gel
Each for $528.50
Add to cart
Description
Description
Can be optimal for lab use and precise Q.C. measurements.
  • 0 to 14 ph range
  • -5 to +100°C range
  • 102mmL, 10mm dia.
Specifications
Specifications
pH Combination Electrode