Fisher Scientific™ Adjustable-Flame Burners
Ideal for very slow heating or quick boiling
 View more versions of this product
Catalog No. 03-962PQ
 N/A
N/A
This item has been discontinued and is no longer available. Please call customer service for assistance: 1-800-766-7000.  View substitute products »
Required Accessories : Tubing. Fisherbrand Natural Rubber Latex Tubing (50 ft. length, Cat. No. 14-178-2D) and Fisherband Pure Natural Rubber Tubing (12 ft. length, 14-150) are recommended.<br /><br />
    • Universal straight-tube burners have flames adjustable from 0.75 to 12 in. (20 to 305mm) Obtain heat output of over 1465w (5000 BTU/hr.)
    • Consume 5 cu. ft./hr. of gas
    • Flame retainer (supplied) prevents flame lifting or extinguishing
    • Adjustable air and gas controls ensure complete combustion
    • Tapered, serrated gas connection
    • Nonferrous, rustproof construction
    • Separate models for natural and LP gases
  • Description & Specifications
Related Products
Product Content Feedback