Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Fisherbrand Adjustable Jacobs Chuck
Accepts stirrer shafts from 1/16 in. to 3/8 in. (1.5 mm to 10mm) in diameter.
Description
- Fits 3/8 in. (10mm) diameter motor shafts
- Accepts stirrer shafts from 1/16 in. to 3/8 in. (1.5 mm to 10mm) in diameter
Specifications
Adjustable Jacobs Chuck | |
10mm dia. Motor Shaft |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title