Learn More
Fisher Scientific™ Air-Powered Analog Overhead Stirrers
Variable speed and explosion-proof stirrers ideal for safely mixing all types of solvents, lacquers and volatile chemicals without the possiblity of spark ignition
 View more versions of this product
Catalog No. 14-500-217
  Check Availability
Add to cart
Specifications
Description
  • Needle valve allows variation in mixing speed
  • Special muffler ensures quieter operation
  • Reversible models have motors which can be set to rotate in either direction by connecting the air supply line to the right port for counter-clockwise rotation or the left port for clockwise rotation
14-500-216 (Model AP50R)
  • High torque, high speed reversible stirrer
  • Includes a 5/8 in. diameter support rod
  • Can be used in ambient temperatures up to 250°F (120°C)
14-500-217 (Model AP20R)
  • High torque, heavy duty reversible stirrer specially designed for use with more viscous materials
  • Comes with a heavy duty gear reduction unit that will extend the life of the stirrer when used in continuous agitation of highly viscous materials
14-500-218 (Model AP20)
  • General purpose stirrers specially designed for use with more viscous materials
  • Constant speed attained through a 7-1 gear reduction thus lowering speed fluctuations due to changes in air pressure
14-500-219 (Model AP10)
  • Light to medium duty stirrers ideal for all types of solvents, lacquers, oils, sythetics and chemicals
  • Can be used in ambient temperatures up to 250°F (120°C)
Using a filter/ regulator with air supply line is recommended
Description & Specifications
Specifications
0.37 in.
6 lb.
0.95cm
0.5
10 L x 0.56 in. dia. support rod, 4 ft. air hose with double fittings for connection to air supply
676.28 oz.
20L
1000rpm
2.7kg
Safely mixing all types of solvents, lacquers and volatile chemicals
30 to 80psi
294
20000mL
10000