Learn More
Fisherbrand™ Alundum™ Bedding Material
Products 2
Catalog Number Description Price Qty Availability  
Catalog Number Description Price Qty Availability  
A620-500
View Documents
N/A Packaging: Amber Glass; Quantity: 500g This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
N/A
A634-3
View Documents
N/A 3killogram This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
N/A
Description
Description
  • Separates steel sample from boat to prevent fusions or chemical reactions during carbon determinations
  • Treated to remove carbonaceous matter and surface alkali
  • Less than 0.0015% carbon content