Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Fisher Science Education™ AX-800 Series Compound Research Microscopes with Koehler Illumination
SCI_ED

Advanced research microscopes for university and laboratory use.

Manufacturer:  Fisher Science Education™ AX813PL

 View more versions of this product

Catalog No. S71016C

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

  • Option between infinity corrected optics or plan phase optics, and choice of binocular or trinocular heads
  • Professional frame with low-position coaxial focusing allows one-handed operation
  • Built-in low-drive large mechanical stage (160mm x 140mm) with extended post for X and Y control
  • 30° inclined ergonomic viewing head
Specifications

Specifications

11350112
Trinocular
Koehler LED with Intensity Control 3w
Abbe/Iris (N.A. 1.25)
Infinity Corrected
Infinity Corrected
15.4 lb.
IEC 110/220 V (UL/CSA)
Compound Research Microscope
N-WF 10X/20 mm with diopter adjustment, +/-5 diopter (Rubber eye shields included)
IEC Power Cord, Built-in Cord Hanger, Dustcover, Allen Key
4X, 10X, 40X, 100X
Reversed quintuple revolving
7 kg
16 x 7.71 x 16.2 in. (407 x 196 x 414 mm)
5 year limited warranty, excluding bulbs, cords, and fuses
SDS
Documents
Product Certifications
Promotions

Promotions

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit