Learn More
Fisherbrand™ Baume Hydrometers
Baume “heavy” scale for working with liquids heavier than water.
Description
Baume “heavy” scale for working with liquids heavier than water.
- Permanently marked with individual serial numbers
Order Info
Use Fisherbrand cylinder 08-530J with 250mL of liquid for submersion, available separately
Certifications
A certificate is provided to indicate traceability to standards provided by NIST™.
Compliance
Accuracy meets or exceeds ASTM™ E 100 specifications, Calibrated according to ASTM E 126 specifications.
Specifications
Baume Hydrometer | |
Without 3-point certification | |
35 to 70° |
0.5° | |
300 mm | |
Baume 'Heavy' Scale For Liquids Heavier than Water |