Learn More
Fisher Science Education™ Digital Tablet Compound Microscope
All-in-one digital microscope fully functional without computer.
Supplier: Fisher Science Education™ BTW1213RLED
Description
- Integrated tablet for simultaneous viewing through traditional eyepiece
- Pre-loaded Motic Apps for teachers, students and researchers to connect, view and share live images
- Built-in camera
- Wifi interface
Specifications
Coarse and Fine | |
LCD | |
LED | |
Compound Microscope | |
Reversed Quadruple revolving |
1.25 NA Abbe adjustable | |
Monocular | |
4X, 10X, 40X | |
USB: 1280 x720/1280x 1024 preview mode, 2592x1944 (5MP) still imageWifi: 1280 x960 preview mode, 1280 x960 (1.3MP) still image |